See the table below for the pore size ranges for each of the Hi-Flow membranes.
Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
10 sheets | Please contact Customer Service for Availability | €156.00 |
About This Item
€156.00
packaging
pack of 10 sheets (2 sheets of each speed)
Quality Segment
manufacturer/tradename
Hi-Flow Plus
backing thickness
4 mil
size
30.5 cm × 25 cm
General description
Application
- Hi-Flow™ Plus membrane is a critical raw material suitable for use in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing and animal health.
- Hi-Flow™ Plus membranes are suitable for use with all lateral flow test techniques including competitive, sandwich, and multiplex detection assays.
- They are optimised for use in both qualitative and quantitative test types and pair well with lateral flow detector particles including colloidal gold nanoparticles, dyed/ fluorescent microspheres etc.
- Hi-Flow™ Plus membranes are suitable for use in lateral flow tests designed to detect analytes present in a broad range of samples matrices including, blood, saliva, urine, nasopharyngeal swabs, sweat and others.
Features and Benefits
- Consistent Flow & Surface Chemistry, Robust manufacturing process
- Proprietary Surface Chemistry Optimised for Lateral Flow Applications
- High Quality Level: MQ400
- Trusted Partner and Brand with Decades of membrane manufacturing experience
- High production capacity, high stock levels, multiple manufacturing lines
- Convenient samples packs available in several formats
- Manufacturing scale quantities available to purchase online with immediate shipping.
- Consistent capillary flow time and membrane thickness
- Superior surface morphology allows for consistent and even flow distribution of reagents across the membrane surface
- Sharp, intense, and uniform test and control line formation
- Excellent intra-lot and inter-lot consistency.
- Better Assay sensitivity
- Excellent reproducibility
- Improved lateral flow test manufacturing yields.
- Quick Delivery and Lead time.
- No supply disruption
- Excellent Technical support
Packaging
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description backing thickness 4 mil, size 30.5 cm × 25 cm, pack of 10 sheets (2 sheets of each speed) | description backing thickness 2 mil, size 30.5 cm × 25 cm, pack of 10 sheets (2 sheets of each speed) | description backing thickness 4 mil (100 μm), filter W × L 30.5 cm × 25 cm, pack of 5 sheets | description backing thickness 4 mil (100 μm), filter W × L 30.5 cm × 25 cm, pack of 5 sheets |
| backing thickness 4 mil | backing thickness 2 mil | backing thickness 4 mil (100 μm) | backing thickness 4 mil (100 μm) |
| Quality Level 400 | Quality Level 400 | Quality Level 400 | Quality Level 400 |
| manufacturer/tradename Hi-Flow Plus | manufacturer/tradename Hi-Flow Plus | manufacturer/tradename Hi-Flow Plus | manufacturer/tradename Hi-Flow Plus |
| packaging pack of 10 sheets (2 sheets of each speed) | packaging pack of 10 sheets (2 sheets of each speed) | packaging pack of 5 sheets | packaging pack of 5 sheets |
| size 30.5 cm × 25 cm | size 30.5 cm × 25 cm | size - | size - |
Still not finding the right product?
Explore all of our products under Hi-Flow™ Plus Variety Sample Pack
signalword
Danger
Storage Class
11 - Combustible Solids
hcodes
Hazard Classifications
Aquatic Chronic 3 - ED ENV 1 - Flam. Sol. 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Hello there, I want to ask what are the pore sizes for the NC membranes you have in the lateral flow assay kit (cat number: HFSAMPLE04). I have 5 membranes in the kit (HF075, HF090, HF120, HF135, HF180)
1 answer

