Key Documents
EZSSGASK1
Millipore
Silicon gasket for EZ-Stream® pump
To replace original gasket
Select a Size
1 EACH
€25.30
€25.30
Estimated to ship on03 March 2025Details
Request more information
Select a Size
Change View
1 EACH
€25.30
About This Item
UNSPSC Code:
41116500
NACRES:
NB.79
€25.30
Estimated to ship on03 March 2025Details
Request more information
feature
repair kit
packaging
pkg of 1 numbers
manufacturer/tradename
EZ-Stream®
application(s)
bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring
compatibility
for use with EZ-Stream®
General description
The silicon gasket is for the replacement of the original gasket that ensures the tightness between the housing and the bottom plate of the EZ-Stream® vacuum pump to prevent liquid to seep inside the pump. The EZ-Stream® vacuum pump is a liquid transfer pump with a compact design that makes the pump ideal for use on both the workbench and in the laminar flow hood
Application
The silicon gasket ensures tightness between the housing and the bottom plate of the EZ-Stream® pump that is used for microbiology analysis of Beer, Dairy, Bottled Water, Pharmaceutical Products Analysis, Soft Drink, Cider, Sports Drink & Wine.
Legal Information
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany