Skip to Content
Merck
All Photos(2)

Documents

EZS0RING2

Millipore

O-Rings for EZ-Stream® Vacuum Pump

Liquid Leakage Barrier

Synonym(s):

O-Ring Seal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
NACRES:
NB.79

material

EPDM rubber O-ring

description

O-ring

feature

repair kit

packaging

pkg of 2 units

manufacturer/tradename

EZ-Stream®

application(s)

bioburden testing
food and beverages
industrial qc
pharmaceutical
water monitoring

compatibility

for use with EZ-Stream® Vacuum Pump

General description

O-Rings for EZ-Stream® Vacuum Pump is designed to create a barrier to any liquid leakage during the filtration process of test samples.
This reference includes two (2) O-rings to replace original O-rings that ensures the tightness between the hose barbs and the liquid pump inside the system.
The EZ-Stream® Vacuum Pump is ideal to be used both on work bench and laminar flow hood, where its small footprint saves laboratory space.

Application

O-Rings for EZ-Stream® Vacuum Pump is used to prevent any leakage of filtration liquid during the filtration process.

Legal Information

EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

related product


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service