Skip to Content
Merck
All Photos(1)

Key Documents

EZFITMAKIT

Millipore

EZ-Fit® Manifold maintenance kit

Including lubricant and brush, for use with EZ-Fit® Filtration Manifold

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€211.00

€211.00


Estimated to ship on30 April 2025Details


Request more information

Select a Size

Change View
1 EA
€211.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

€211.00


Estimated to ship on30 April 2025Details


Request more information

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
environmental
food and beverages
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

General description

EZ-Fit® Manifold maintenance kit helps to clean and decontaminate the EZ-Fit® Manifold regularly depending upon the schedule of use and type of samples filtered. The kit contains nonwoven wipes to clean the filtration head, the valve, valve button, and o-ring, using the recommended cleaning agents. A dedicated small brush provided in the kit is used to clean the small canal in the filtration head support and a big brush to clean the valve housing. The kit is also provided with lubricant to lubricate the o-rings.

Application

The EZ-Fit® filtration Manifold is used for universal laboratory filtrations and, the maintenance kit helps to clean and decontaminate the EZ-Fit® Manifold regularly.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service