Skip to Content
Merck
All Photos(1)

Key Documents

EZFITFRIT3

Millipore

EZ-Fit® Manifold removable porous filter support

stainless steel, for use with EZ-Fit® Filtration Manifold

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
€93.10

€93.10


Available to ship on16 April 2025Details


Request more information

Select a Size

Change View
3 EA
€93.10

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

€93.10


Available to ship on16 April 2025Details


Request more information

material

stainless steel

Quality Level

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 3 units

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

General description

EZ-Fit® Manifold porous filter support is a spare accessory of Microfil® filtration head. It can be removed simply by pressing down anywhere around its edge. The Microfil® filtration head is designed to improve ease of use, reduce maintenance, and limit the risk of contamination.

Application

EZ-Fit® Manifold porous filter support is a spare accessory of Microfil® filtration head used during filtration of the test samples for general microbiological analysis, bioburden testing, and water monitoring.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. For this catalogue EZFITFRIT3 wich is the pore size? I want to know if is possible that a gelatin solution (10%) can pass through the filter soport.

    1 answer
    1. The item number EZFITMIHE3 corresponds to the Microfil filtration head for EZ-Fit Manifold. This manifold head accommodates filtration membranes with a diameter of 47mm.
      The removable porous filter support in the manifold head, EZFITFRIT3, have a diameter of 38.4mm. These frits are only used as a filtration support on which you place the membranes with a defined very small pore-size for microbes retention. The frits as such has no specified pore-size.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service