Skip to Content
Merck
All Photos(2)

Key Documents

EZFITHIHE

Millipore

EZ-Fit® Manifold Filtration Head

Rubber Stopper Filtration Head (Standard No. 8), for use with EZ-Fit® Filtration Manifold

Synonym(s):

For bioburden application, Rubber Stopper Filtration Head (Standard No. 8), filtration head for manifold

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€279.00
3 EA
€809.00

€279.00


Available to ship on11 April 2025Details


Request more information

Select a Size

Change View
1 EA
€279.00
3 EA
€809.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

€279.00


Available to ship on11 April 2025Details


Request more information

material

316L stainless steel

Quality Level

Sterilization Compatibility

autoclavable compatible

feature

Rubber Stopper Filtration Head (Standard No. 8)

packaging

pkg of 1 unit (EZFITHIHE1)
pkg of 3 units (EZFITHIHE3)

manufacturer/tradename

EZ-Fit®

input

sample type beer
sample type bottled water
sample type pharmaceutical(s)
sample type cosmetics

application(s)

bioburden testing
cosmetics
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The filtration head has been designed to improve ease-of-use, reduce maintenance, and limit the risk of contamination. The Standard No. 8 perforated stopper can be used with steel or glass funnels

Application

General microbiological analysis for presence of particulate contamination - Beer; Bottled Water; Product Analysis; Environmental Analysis; Bioburden Testing; Water Monitoring

Packaging

EZFITHIHE1: 1 unit
EZFITHIHE3: 3 units
comes without Rubber Stopper n°8

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service