Skip to Content
Merck
All Photos(1)

Key Documents

EZFITH0LD1

Millipore

EZ-Fit® Manifold, 1-place for rubber stopper tulip

1-place, for rubber stopper tulip

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€1,960.00

€1,960.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
€1,960.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

€1,960.00


Please contact Customer Service for Availability

Request more information

material

stainless steel

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 unit (1x EZFITBASE1
1x EZFITHIHE1)

manufacturer/tradename

EZ-Fit®

input

sample type beer
sample type bottled water
sample type pharmaceutical(s)
sample type cosmetics

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

compatibility

for rubber stopper tulip

shipped in

ambient

General description

EZ-Fit® Manifold′s unique design optimizes workflow and maintenance and reduces the risk of contamination. Different filtration heads fit both reusable and disposable filtration devices. Low height design increases comfort when used in laminar flow hoods. EZ-Fit® Manifold prevents biofilm formation because every single component can be removed for easy cleaning and autoclaving.

Application

The EZ-Fit® Manifold is a filtration manifold used for universal laboratory filtrations. It supports the simultaneous filtration of one test sample. The manifold can be used with EZ-Fit® filtration units, Microfil® filtration system, a 55-Monitor Plus, and standard glass or stainless-steel funnels.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service