Skip to Content
Merck
All Photos(1)

Key Documents

ET-TMB-M

Sigma-Aldrich

Stop Solution

Sign Into View Organizational & Contract Pricing

Select a Size

ET-TMB
€34.00

€34.00


Estimated to ship on12 February 2025Details


Request a Bulk Order

Select a Size

Change View
ET-TMB
€34.00

About This Item

UNSPSC Code:
12161703
NACRES:
NA.84

€34.00


Estimated to ship on12 February 2025Details


Request a Bulk Order

Quality Level

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service