Skip to Content
Merck
All Photos(1)

Key Documents

DX0796

Supelco

1,2-Dichloroethane

OmniSolv®

Synonym(s):

Dichloroethane, Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein:
605264
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-458-1
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
83 °C (lit.)
vapor pressure:
87 mmHg ( 25 °C)

vapor density

3.4 (20 °C, vs air)

Quality Level

vapor pressure

87 mmHg ( 25 °C)

product line

OmniSolv®

Assay

≥99% (GC)

form

liquid

autoignition temp.

775 °F

expl. lim.

15.6 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.02% Water (H2O)
≤0.3 μeq/g Titrable acid

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

transition temp

flash point 13 °C

density

1.25 g/cm3 at 20  °C
1.256 g/mL at 25 °C (lit.)

UV cutoff

≤230 nm

UV absorption

λ: 230 nm Amax: ≤1.00
λ: 240 nm Amax: ≤0.30
λ: 250 nm Amax: ≤0.10
λ: 300 nm Amax: ≤0.005
λ: 400 nm Amax: ≤0.005

shipped in

ambient

storage temp.

room temp

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DX0796P-1 is packaged in poly coated glass bottles
OmniSolv®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.0% minColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)500ppt maxFormClear liquidFree halogensTo pass testInfrared SpectrumConformsTitrable acid0.3µeq/g maxUV Abs. at 230 nm1.00AU maxUV Abs. at 240 nm0.30AU maxUV Abs. at 250 nm0.10AU maxUV Abs. at 300 nm0.005AU maxUV Abs. at 400 nm0.005AU maxUV Cut-off230nm maxWater (H2O)0.02% max
Evaporation Number: 4.1

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service