Skip to Content
Merck
All Photos(2)

Key Documents

414913

Sigma-Aldrich

Iron(II) ethylenediammonium sulfate tetrahydrate

98%

Synonym(s):

Ferrous ethylenediammonium sulfate tetrahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€69.90
100 G
€151.00

€69.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€69.90
100 G
€151.00

About This Item

Linear Formula:
FeSO4 · NH3CH2CH2NH3SO4 · 4H2O
CAS Number:
Molecular Weight:
382.15
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€69.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

reaction suitability

core: iron
reagent type: catalyst

SMILES string

O.O.O.O.[Fe++].NCCN.OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/C2H8N2.Fe.2H2O4S.4H2O/c3-1-2-4;;2*1-5(2,3)4;;;;/h1-4H2;;2*(H2,1,2,3,4);4*1H2/q;+2;;;;;;/p-2

InChI key

GAGUCKQNZZFVCU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Iron(II) ethylenediammonium sulfate tetrahydrate is an iron precursor used in the synthesis of electrocatalysts. [1]

Application

Iron(II) ethylenediammonium sulfate tetrahydrate can be used as an iron precursor salt for the synthesis of Fe/N/carbon-based electrocatalysts for the oxygen reduction reaction.[2] It is also used to functionalize multi-wall carbon nanotubes (MWCNTs) with maghemite nanocrystals to enhance the magnetic and electronic properties of MWCNTs. These functionalized nanocomposites are applied for the decontamination of pollutants in water.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of the pyrolysis atmosphere and nature of iron precursor on the structure and activity of Fe/N based electrocatalysts for the oxygen reduction reaction
Dominguez C, et al.
International Journal of Hydrogen Energy, 41(47), 22560-22569 (2016)
Synthesis and characterization of maghemite nanocrystals decorated multi-wall carbon nanotubes for methylene blue dye removal
Bhakta AK, et al.
Journal of materials science & technology, 54(1), 200-216 (2019)
Effect of the pyrolysis atmosphere and nature of iron precursor on the structure and activity of Fe/N based electrocatalysts for the oxygen reduction reaction
Domnguez, Carlota and Pea
International Journal of Hydrogen Energy, 41, 22560-22569 (2016)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service