Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 vial | Estimated to ship on13 July 2026fromMerck Life Science BV/SRL | €194.00 |
About This Item
€194.00
Estimated to ship on13 July 2026Details
form
liquid (Reagent)
Quality Segment
packaging
vial of 600 μL
manufacturer/tradename
Millipore
growth mode
N/A
technique(s)
Northern blotting: suitable, Southern blotting: suitable
shipped in
dry ice
storage temp.
−20°C
Features and Benefits
Preparation Note
Other Notes
The DNA includes fragments ranging from 100-3,000 base pairs. The 500 and 1,500 base pair bands have increased intensity to serve as reference points. Novel Green fluorescence dye was added for producing an instantvisualization of DNA bands upon illuminating with UV or Blue Light.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) Northern blotting: suitable, Southern blotting: suitable | technique(s) Northern blotting: suitable, Southern blotting: suitable | technique(s) Northern blotting: suitable, Southern blotting: suitable | technique(s) Northern blotting: suitable, Southern blotting: suitable |
| form liquid (Reagent) | form - | form liquid (Reagent) | form liquid (Reagent) |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



