Skip to Content
Merck
All Photos(1)

Key Documents

DPLUS-1KS

Sigma-Aldrich

DirectLoad Plus 1kb DNA Ladder with fluorescent DNA stains

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€186.00

€186.00


Estimated to ship on19 February 2025Details



Select a Size

Change View
1 VIAL
€186.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.52

€186.00


Estimated to ship on19 February 2025Details


packaging

vial of 600 μL

Quality Level

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

Northern blotting: suitable
Southern blotting: suitable

shipped in

dry ice

storage temp.

−20°C

Features and Benefits

It′s suitable for use as an electrophoresis marker for DNA

Storage and Stability

-20 °C for up to 24 months, or 4 °C up to 12 months or room temperature for up to six months.

Other Notes

DirectLoad Plus 1kb DNA Ladder with fluorescent DNA stains contains 12 fragments, which are obtained from PCR products and double-stranded DNA digested with appropriate restriction enzymes equilibrated to 10 mM Tris-HCl (pH 8.0) and 10 mM EDTA after phenol extraction.
The DNA includes fragments ranging from 100-10,000 base pairs. The 1,000 and 3,000 base pair bands have increased intensity to serve as reference points. Novel Green fluorescence dye was added for producing an instantvisualization of DNA bands upon illuminating with UV or Blue Light.

Legal Information

DirectLoad is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service