C5VP080A
Cellicon® Filter Assembly with Durapore® Membrane
filtration area 0.8 m2
Synonym(s):
Cellicon® Filter Assembly with Durapore® Membrane
About This Item
Recommended Products
material
PVDF membrane
polyester screen
polypropylene housing (polypropylene silicone)
silicone O-ring
sterility
non-sterile; γ-irradiated
storage condition
dry at room temperature
room temperature (15-30°C)
parameter
7.15 L/min-m2 flow rate
technique(s)
tangential flow filtration (TFF): suitable
D × W × H
12.2 cm (4.8 in) × 23.4 cm (9.2 in) × 37.6 cm (14.8 in)
filtration area
0.8 m2
volume
0.73 L , Filter Only
1.1 L , Assembly
1100 mL , Assembly
730 mL
matrix
Durapore®
pore size
5.0 μm
pH
4-8
application(s)
mAb
recombinant protein
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Predictable linear scalability from lab to manufacturing scale
- Reliable and reproducible performance
- Gentle on cells, maximizing cell density and viability
- Easy installation, with smooth connection to bioreactor through welding or sterile connectors
Packaging
Other Notes
Consolidation of product specific testing, quality and regulatory information to simplify compliance requirements.
Convenient 24/7 access to up-to-date product information.
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Enhance perfused seed train efficiency with our cell retention solution. Real-time monitoring and control.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service