BX1657
Buffer pH 8.0 Clear
Synonym(s):
buffer solutions
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
Recommended Products
grade
analytical standard
Quality Level
Agency
traceable to NIST
application(s)
environmental
food and beverages
general analytical
industrial qc
pharmaceutical
format
mixture
shipped in
ambient
storage temp.
15-25°C
Application
This pH 8.0 buffer solution can be used in routine analytical processes for the calibration of pH meters and maintaining the pH of solutions.
Features and Benefits
- Traceable to NIST standards
- A ready-to-use solution available in a sealed bottle with convenient packaging to protect against contamination
- Accompanied with a certificate of analysis (CoA) for an effective audit
- Accurate up to +/- 0.02 at 25 °C
- The expiry date of the product can be found in the CoA
- Calculated uncertainty value is for the duration of transportation as well as storage
Specifications
Tolerance: ±0.02
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service