Skip to Content
Merck
All Photos(2)

Key Documents

ABN645

Sigma-Aldrich

Anti-RIM 2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Regulating synaptic membrane exocytosis protein 2, Rab-3-interacting molecule 2, RIM 2, Rab-3-interacting protein 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€370.00

€370.00


Estimated to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Estimated to ship on14 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RIMS2(9699)

General description

Rab3-interacting molecules (RIMs) are synaptic proteins necessary for neuronal transmission and plasticity. RIM 1 and RIM 2 proteins are expressed in overlapping but distinct patterns throughout the brain. While the ablation of either gene was not lethal in mice, the deletion of both resulted in postnatal mortality. This lethality is due to a defect in neurotransmitter release; synapses without RIM proteins can still release neurotransmitters but are unable to do so in response to normal Ca2+ triggers. Like RIM 1, RIM 2 is thought to be an effector protein for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner.

Specificity

This antibody may recognize many isoforms and is predicted to have no cross-reactivity to other RIM proteins.

Immunogen

Epitope: center region
KLH-conjugated synthetic peptide corresponding to the center region of human RIM 2.

Application

Anti-RIM 2 Antibody is an antibody against RIM 2 for use in western blotting, IHC & immunofluorescence.
Immunohistochemistry Analysis: 5 μg/mL from a representative lot detected RIM 2 in human brain tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected RIM 2 in human brain cells.



Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in rat brain tissue lysate with and without blocking peptide.

Western Blotting Analysis: 1 μg/mL of this antibody detected RIM 2 in rat brain tissue lysate with and without blocking peptide.

Target description

~86 kDa observed. Uniprot describes 3 isoforms produced by alternative splicing ranging between ~31 kDa and ~160 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service