Skip to Content
Merck
All Photos(1)

Key Documents

BI0166

Sigma-Aldrich

Activator Reagent 0.3M BMT in AcN

Biosynthesis Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€1,530.00

€1,530.00


Estimated to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
€1,530.00

About This Item

UNSPSC Code:
41116006

€1,530.00


Estimated to ship on14 April 2025Details


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Coupling Reactions
reagent type: catalyst

transition temp

flash point 2 °C (Acetonitrile)

shipped in

ambient

storage temp.

2-30°C

General description

Activator Reagent 0.3M 5-benzylmercaptotetrazole (BMT) in acetonitrile (AcN) is also known as benzylthiotetrazole (BTT) or benzyl-1H-tetrazole). It is used as a phosphoramidite activator reagent for oligonucleotide synthesis.

Application

0.3M BMT in AcN can be used as a reagent in solid phase phosphorylation of the oligomers. The activator reacts with the amidite group to form a highly reactive intermediate. The intermediate then forms an internucleotide bond with the 5′hydroxyl group of the growing oligonucleotide chain. The high activation potential of this reagent may be due to its greater acidity (pKa=4.08), which makes it easier to protonate the trivalent phosphorus in the initial stage of activation and increase the speed of the coupling reaction.

Features and Benefits

  • efficient and rapid activation reagent that produces less
  • phosphoramidite waste      
  • higher solubility at lower temperatures      
  • does not crystallize or clog lines       
  • safer reagent to use

Analysis Note

Color (APHA): 10APHA max
Component: Acetonitrile Purity (By GC): 99.9% min
Form: Clear Liquid
Identity (IR-spectrum): Conforms
Molarity of 5-Benzylmercaptotetrazole: 0.28-0.32M
Water (H2O): 1-30ppm

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup - (Acetonitrile)

Flash Point(C)

2 °C - closed cup - (Acetonitrile)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service