Skip to Content
Merck
All Photos(1)

Key Documents

935409

Sigma-Aldrich

Sulfo-NHS-LC-LC-Biotin

≥95%

Synonym(s):

3-Pyrrolidinesulfonic acid, 1-[[6-[[6-[[5-(hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl)-1-oxopentyl]amino]-1-oxohexyl]amino]-1-oxohexyl]oxy]-2,5-dioxo-, monosodium salt, [3aS-(3aα,4β,6aα)]-[partial]-, EZ-Link Sulfo-NHS-LC-LC-Biotin, Hexanoic acid, 6-[[6-[[5-[(3aS,4S,6aR)-hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]-1-oxohexyl]amino]-, 2,5-dioxo-3-sulfo-1-pyrrolidinyl ester, sodium salt (1:1), Sulfo-NHS-XX-Biotin

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€149.00
50 MG
€298.00

€149.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€149.00
50 MG
€298.00

About This Item

Empirical Formula (Hill Notation):
C26H40N5O10S2·Na
CAS Number:
Molecular Weight:
669.74
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

€149.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95%

form

solid

storage temp.

−20°C

SMILES string

[Na].O=C1NC2CSC(CCCCC(=O)NCCCCCC(=O)NCCCCCC(=O)ON3C(=O)CC(C3=O)S(=O)(=O)O)C2N1

InChI

InChI=1S/C26H41N5O10S2.Na/c32-20(27-14-8-2-4-12-23(35)41-31-22(34)15-19(25(31)36)43(38,39)40)10-3-1-7-13-28-21(33)11-6-5-9-18-24-17(16-42-18)29-26(37)30-24;/h17-19,24H,1-16H2,(H,27,32)(H,28,33)(H2,29,30,37)(H,38,39,40);/t17-,18-,19?,24-;/m0./s1

InChI key

RWNPTJCDHAXLGI-BJPAGVOZSA-N

Application

Sulfo-NHS-LC-LC-biotin finds extensive applications in scientific research for the purpose of protein labeling and detection. Its utilization in protein purification allows for the isolation of biotinylated proteins from intricate mixtures. Moreover, it plays a vital role in protein-protein interaction studies, enabling investigations into the binding affinity and specificity of proteins. Additionally, it serves as a valuable tool in protein quantification, facilitating the measurement of specific protein levels within a given sample.

Features and Benefits

Sulfo-NHS-LC-LC-biotin is a frequently employed chemical compound in scientific investigations aimed at examining and characterizing proteins. This compound serves as a biotinylation agent, allowing for the labeling of proteins in diverse applications, including protein purification, studies on protein-protein interactions, and protein quantification. Sulfo-NHS-LC-LC-biotin works by reacting with the primary amines of proteins to form stable amide bonds. This results in the biotinylation of the protein, which allows for the detection and isolation of the protein using streptavidin or avidin conjugated to a detection system, such as a fluorescent or radioactive label.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A reagent for covalently attaching biotin to proteins via a cleavable connector arm.
C A Mouton et al.
Archives of biochemistry and biophysics, 218(1), 101-108 (1982-10-01)
D Castelletti et al.
Clinical and experimental immunology, 136(2), 365-372 (2004-04-17)
Hodgkin's lymphoma patients treated with an anti-CD25 Ricin toxin A-chain (RTA)-based Immunotoxin (RFT5.dgA) develop an immune response against the toxic moiety of the immunoconjugate. The anti-RTA antibody response of 15 patients showing different clinical features and receiving different total amounts
M Ichikawa et al.
Bioorganic & medicinal chemistry letters, 11(13), 1769-1773 (2001-06-27)
We have prepared several mechanism-based affinity-labeling agents for possible use in isolating N-acetylglucosaminidase, in which an N-acetylglucosamine is linked to an o-monofluoro- or difluoro-methyl phenoxy glycoside with or without a cleavable disulfide group in the tether to biotin.
Chemical probes of extended biological structures: synthesis and properties of the cleavable protein cross-linking reagent [35S]dithiobis(succinimidyl propionate).
A J Lomant et al.
Journal of molecular biology, 104(1), 243-261 (1976-06-14)

Questions

  1. Whats the protocol used for biotinylation using this reagent (935409)?

    1 answer
    1. The method of biotinylating is considered proprietary. However, a general reference regarding the biotinylation of proteins is listed in the 'PEER REVIEWED PAPERS' section of the product page:

      A reagent for covalently attaching biotin to proteins via a cleavable connector arm.
      C A Mouton et al.
      Archives of biochemistry and biophysics, 218(1), 101-108 (1982-10-01)

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service