Skip to Content
Merck
All Photos(1)

Key Documents

AX1235

Supelco

Ammonium Bifluoride

Practical

Synonym(s):

Ammonium Bifluoride, Ammonium Hydrogen Fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H5F2N
CAS Number:
Molecular Weight:
57.04
MDL number:
UNSPSC Code:
12352301
EC Index Number:
215-676-4
grade:
for analytical purposes
form:
flakes
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

vapor pressure

1 hPa ( 20 °C)

Assay

≥96.0%

form

flakes

color

white, typical

pH

2 (20 °C, 5.7 g/L in H2O)

bp

238 °C/1013 hPa (decomposition)

mp

126 °C

shipped in

ambient

storage temp.

room temp

SMILES string

F.F.N

InChI

1S/2FH.H3N/h2*1H;1H3

InChI key

KVBCYCWRDBDGBG-UHFFFAOYSA-N

Related Categories

Application


  • Enhanced removal of toxic Cr(vi) and Pb(ii) from water using carboxylic terminated Ti(3)C(2)T(x) nanosheets.: This study evaluates the efficiency of Ti(3)C(2)T(x) nanosheets, enhanced with carboxylic groups, in removing toxic chromium and lead from water, demonstrating a potential application for ammonium bifluoride in environmental remediation (Shah et al., 2023).

  • Low-temperature method for desiliconization of polymetallic slags by ammonium bifluoride solution.: This research presents a low-temperature approach using ammonium bifluoride for desiliconizing polymetallic slags, offering a significant improvement in processing materials in metallurgy (Smorokov et al., 2023).

  • Rapid decomposition of geological samples by ammonium bifluoride (NH(4)HF(2)) for combined Hf-Nd-Sr isotope analyses.: The study introduces a method for the rapid decomposition of geological samples using ammonium bifluoride, enhancing the efficiency of isotopic analysis in geoscience (Újvári et al., 2021).

  • Innovative high-temperature ammonium bifluoride fusion and rapid analysis of elements with nuclear forensic value.: This paper discusses an innovative method for the high-temperature fusion of ammonium bifluoride to quickly analyze elements critical for nuclear forensics, demonstrating its significant application in security and environmental monitoring (Bradley et al., 2021).

  • Sterically Encumbered Tris(trialkylsilyl) Phosphine Precursors for Quantum Dot Synthesis.: This research explores the synthesis of quantum dots using tris(trialkylsilyl) phosphine precursors facilitated by ammonium bifluoride, highlighting its role in advanced materials engineering (Chandrasiri et al., 2020).

Analysis Note

Assay: 96% min
Color: White (typical)
Form: Flakes

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service