Skip to Content
Merck
All Photos(1)

Key Documents

1.01160

Supelco

Ammonium hydrogen difluoride

pure

Synonym(s):

Ammonium bifluoride, Etching powder

Sign Into View Organizational & Contract Pricing

Select a Size

5 KG
€191.00

€191.00


Estimated to ship on28 March 2025Details


Request a Bulk Order

Select a Size

Change View
5 KG
€191.00

About This Item

Linear Formula:
NH4HF2
CAS Number:
Molecular Weight:
57.04
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.21
grade:
for analytical purposes

€191.00


Estimated to ship on28 March 2025Details


Request a Bulk Order

grade

for analytical purposes

Quality Level

vapor pressure

1 hPa ( 20 °C)

pH

2 (20 °C, 5.7 g/L in H2O)

bp

>238 °C/1013 hPa (decomposition)

mp

125 °C (lit.)

SMILES string

N.F.F

InChI

1S/2FH.H3N/h2*1H;1H3

InChI key

KVBCYCWRDBDGBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service