Skip to Content
Merck
All Photos(1)

Key Documents

ALIGNED25

Millipore

Millipore Alignment Guide for Vacuum Filtration

25 mm, 5/PK

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
€120.00

€120.00


Estimated to ship on29 September 2025Details



Select a Size

Change View
5 EA
€120.00

About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

€120.00


Estimated to ship on29 September 2025Details


material

silicone

technique(s)

vacuum filtration: suitable

diam.

25 mm

application(s)

filtration

shipped in

ambient

General description

Millipore Glass Vacuum Filtration Accessory Product
Alignment Guide, 25 mm, 5/PK

These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.

This item is an accessory part for the Millipore 25 mm Glass Microanalysis Filter Holder Kits.

Application

25 mm, 5/PK

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service