Skip to Content
Merck
All Photos(1)

Key Documents

ACR5000NT

Millipore

Ni-NTA Reagent Kit

suitable for protein purification

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NA.56

product line

Amicon®

technique(s)

protein purification: suitable (His-tagged proteins)

shipped in

wet ice

General description

For use with the Amicon Pro Affinity Conc: 3 ml HisBind Ni-NTA resin, 25ml Bind Buffer (4X), 25 ml Wash buffer (4X), 5ml elution buffer (4X)
Ni-NTA Reagent Kit for His-tagged protein purification.

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

Flash Point(F)

95.9 °F

Flash Point(C)

35.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Protein purification is simpler with Amicon Pro. The device delivers consistent sample preparation for more reliable recovery, uncompromised purity and easier data generation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service