Skip to Content
Merck
All Photos(1)

Documents

70691

Millipore

Ni-NTA His•Bind® Superflow Resin

Synonym(s):

Ni-NTA Resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

do not freeze

application(s)

life science and biopharma

shipped in

ambient

storage temp.

2-8°C

General description

Ni-NTA His•Bind Superflow Resin is a medium-pressure compatible version of the Ni-NTA His•Bind Resin (Cat. No. 70666). It has all the same characteristics, except the agarose matrix has a higher level of crosslinking for higher bead rigidity. Ni-NTA His•Bind Superflow Resin has a binding capacity of 5–50 mg His•Tag fusion protein per ml resin and is compatible with FPLC or gravity-flow applications.

Warning

Toxicity: Flammable (J)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

HIS-BIND is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Superflow is a trademark of Sterogene Bioseparations Inc.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service