Skip to Content
Merck
All Photos(6)

Key Documents

ABT288

Sigma-Aldrich

Anti-PICALM

from rabbit, purified by affinity chromatography

Synonym(s):

Phosphatidylinositol-binding clathrin assembly protein, Clathrin assembly lymphoid myeloid leukemia protein

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€317.00

€317.00


Estimated to ship on28 May 2025Details


Request a Bulk Order

Select a Size

Change View
200 μL
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€317.00


Estimated to ship on28 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PICALM(8301)

General description

Phosphatidylinositol-binding clathrin assembly protein (UniProt: Q13492; also known as PICALM, Clathrin assembly lymphoid myeloid leukemia protein) is encoded by the PICALM (also known as CALM) gene (Gene ID: 8301) in human. PICALM is ubiquitously expressed protein that co-localizes with clathrin in the Golgi area. Five isoforms of PICALM have been reported that are produced by alternative splicing. PICALM acts as an assembly protein that recruits clathrin and adapter protein complex 2 (AP2) to cell membranes at sites of coated-pit formation and clathrin-vesicle assembly. It may also play a role in determining the amount of membrane to be recycled, possibly by regulating the size of the clathrin cage. It is reported to be involved in AP2-dependent clathrin-mediated endocytosis at the neuromuscular junction. PICALM variants are associated with Alzheimer′s disease and are thought to be involved in the endocytosis and recycling of the amyloid protein precursor and it can affect Alzheimer s disease disk primarily by modulating production, transportation, and clearance of beta-amyloid peptides. (Ref.: Xu, W., et al. (2015). Mol Neurobiol. 52(1);399-413).

Specificity

This rabbit polyclonal antibody detects Phosphatidylinositol-binding clathrin assembly protein (PICALM) in human cells. it targets an epitope within 16 amino acids from the C-terminal half. It recognizes all 5 isoforms of PICALM.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal half of human Phosphatidylinositol-binding clathrin assembly protein (PICALM).

Application

Anti-PICALM Antibody, Cat. No. ABT288, is a rabbit polyclonal ntibody that detects PICALM in nhuman cells and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Cell Structure
Western Blotting Analysis: A 1:500 dilution from a representative lot detected PICALM in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PICALM in MOG-G-UVW human astrocytoma cell line (Courtesy of Dr Rhian Thomas from Cardiff University).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot PICALM in Mog-G-UVW human astrocytoma cell lysate and HeLa cell lysate (Courtesy of Dr Rhian Thomas from Cardiff University).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PICALM in Human H4 neuroglioma cell lysate and SH-SY5Y neuroblastoma cell lysate (Courtesy of Dr Rhian Thomas from Cardiff University).

Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected PICALM in human colon and human kidney tissue sections.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected PICALM in HeLa cell lysate.

Target description

~61 kDa observed; 70.76, 68.76, 66.39, 59.92, and 70.04 kDa calculated, respectively for isoforms 1 to 5. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service