Skip to Content
Merck
All Photos(1)

Key Documents

ABT1382

Sigma-Aldrich

Anti-Neurocan

from rabbit

Synonym(s):

Neurocan core protein, 245 kDa early postnatal core glycoprotein, Chondroitin sulfate proteoglycan 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€330.00

€330.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€330.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€330.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

packaging

antibody small pack of 25 μL

technique(s)

immunofluorescence: suitable

isotype

IgG

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

rat ... Ncan(58982)

General description

Neurocan core protein (UniProt: P55067; also known as 245 kDa early postnatal core glycoprotein, Chondroitin sulfate proteoglycan 3) is encoded by the Ncan (also known as Cspg3) gene in rat. Neurocan is a secreted protein of the aggrecan/versican proteoglycan family that is found in early postnatal and adult brain. During early postnatal development neurocan accounts for at least 20% of the protein of the soluble brain proteoglycans. Neurocan mostly contains chondroitin sulfate, but also has N-linked and O-linked oligosaccharides. It is shown to be involved in the modulation of cell adhesion and migration and regulates neurite outgrowth. Neurocan interact with several cell surface molecules, such as N-CAM, L1/Ng-CAM, etc., and modulates the cell-binding and neurite outgrowth promoting activates of these molecules. Neurocan is synthesized with a 22 aa signal peptide that is subsequently cleaved to produce the mature form. Neurocan contains one C-type lectin domain, two EGF-like domains, one Ig-like V type domain, two link domains and one Sushi (CCP/SCR) domain. (Ref.: Rauch, U., et al. (2001) Cell Mol. Life Sci. 58(12-13):1842-56; Zhou, XH et al. (2001). Mol. Cell Biol. 21(17): 5970-78).

Specificity

This rabbit polyclonal antibody detects Neurocan in fetal and adult rat brain.

Immunogen

Epitope: unknown
Full-length Neurocan isolated from 7-day embryonic rat brain and purified by monoclonal antibody immunoaffinity chromatography.

Application

Research Category
Cell Structure
This rabbit polyclonal Anti-Neurocan, Cat. No. ABT1382, is a rabbit polyclonal antibody that detects Neurocan core protein and has been tested for use in Immunofluorescence.

Quality

Evaluated by Immunofluorescence in E14 embryonic rat brain (frozen sections) and adult rat brain (frozen sections).

Immunofluorescence Analysis: A 1:500 dilution of this antibody detected Neurocan in E14 embryonic rat brain (frozen sections) and rat E14 neocortex.

Target description

135.55 kDa calculated.

Physical form

Format: Unpurified
Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo-Yu Wang et al.
Neural regeneration research, 16(8), 1638-1644 (2021-01-13)
Cell transplantation is a potential treatment for spinal cord injury. Olfactory ensheathing cells (OECs) play an active role in the repair of spinal cord injury as a result of the dual characteristics of astrocytes and Schwann cells. However, the specific

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service