Skip to Content
Merck
All Photos(1)

Key Documents

ABE2902

Sigma-Aldrich

Anti-Cdc45

from rabbit

Synonym(s):

DNA replication protein Cdc45

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€311.00

€311.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€311.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€311.00


Estimated to ship on04 June 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, hamster, human

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

hamster ... Cdc45(100689362)
human ... CDC45(8318)
mouse ... Cdc45(12544)

General description

DNA replication protein Cdc45 (UniProt: Q4U3P7; also known as Cdc45) is encoded by the CDC45 gene (Gene ID: 100689362) in hamster. Cdc45 is a replication initiation protein that plays an essential role in the establishment and progression of the replication fork. It is required for formation of pre-initiation complexes at the G1/S transition and for activation of replication origins. Cdc45 associates with the heterohexameric Mcm2-7 helicase and the GINS complex (go-ichi-nii-san, Japanese for 5-1-2-3, for the subunits Sld5, Psf1, Psf2 and Pfs3) to form the CMG (Cdc45-Mcm2-7-GINS) complex. CMG represents the eukaryotic replicative DNA helicase that unwinds origins of replication and drives the replication fork. Cdc45 is also reported to interact with several other replication proteins, including DNA helicase B (DHB), Mcm10, Orc2, TopPB1, and DNA polymerases alpha, delta and epsilon. Expression of Cdc45 is associated with the proliferative status of the cell or tissue. Terminally differentiated and senescent cells do not display Cdc45 activity whereas highly proliferative cells express high levels of Cdc45.

Specificity

This rabbit polyclonal antibody detects Cdc45 in Human and Murine cells.

Immunogen

His-tagged full length hamster recombinant Cdc45.

Application

Anti-Cdc45, Cat. No. ABE2902, is a highly specific rabbit polyclonal antibody that targets DNA replication protein Cdc45 and has been tested for use in Western Blotting.
Western Blotting Analysis: A 1:500 dilution from a representative lot detected Cdc45 in 10 µg of HeLa cell lysate.

Western Blotting Analysis: A representative lot detected Cdc45 in asynchronous, go synchronized and G1 released CHO cells, immunoprecipitated CDC45 from CHO cells, HeLa and NIH3T3 (Wong, P.G., et. al. (2011). PLoS One. 6(3):e17533).

Quality

Evaluated by Western Blotting in CHO cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Cdc45 in 10 µg of CHO cell lysate.

Target description

~65 kDa observed; 65.36 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service