Skip to Content
Merck
All Photos(3)

Key Documents

ABE2889

Sigma-Aldrich

Anti-phospho 4E-BP1 (Ser83)

serum, from rabbit

Synonym(s):

Eukaryotic translation initiation factor 4E-binding protein 1, eIF4E-binding protein 1, Phosphorylated heat- and acid-stable protein regulated by insulin 1, PHAS-I

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€317.00

€317.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€317.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer83)

Gene Information

human ... EIF4EBP1(1978)

General description

Eukaryotic translation initiation factor 4E-binding protein 1 (UniProt: Q13541; also known as 4E-BP1, eIF4E-binding protein 1, Phosphorylated heat- and acid-stable protein regulated by insulin 1, PHAS-I) is encoded by the EIF4EBP1 gene (Gene ID: 1978) in human. 4E-BP1 is a repressor of translation initiation that regulates EIF4E activity by preventing its assembly into the eIF4F complex. It mediates the regulation of protein translation by hormones, growth factors and other stimuli that signal through the MAP kinase and mTORC1 pathways. 4E-BP1 is phosphorylated on serine and threonine residues in response to insulin, EGF, and PDGF. Non-phosphorylated 4E-BP1 bind strongly to eIF4E; however, phosphorylated 4E-BP1 does not bind to eIF4E. 4E-BP1 is reported to act as an inhibitory factor of tumor cells survival and proliferation because of its role as a negative regulator of cap-dependent translation. Although 4E-BP1 serves as a well-characterized substrate of mTORC1, an mTOR-independent phosphorylation of 4E-BP1 has been also been suggested to be associated with the mTOR kinase inhibitor resistance in cancer cells. (Ref.: Shang, ZF et al (2012). Cell Cycle 11(18): 3463 3471).

Specificity

This rabbit polyclonal antibody targets the Eukaryotic translation initiation factor 4E-binding protein 1. It targets an epitope within 13 amino acids surrounding phospho serine 83.

Immunogen

A linear peptide corresponding to 13 amino acids surrounding phospho serine 83 from human Eukaryotic translation initiation factor 4E-binding protein 1 (4E-BP1).
Epitope: unknown

Application

Anti-phospho 4E-BP1 (Ser83), Cat. No. ABE2889, is a highly specific rabbit polyclonal antibody that targets Eukaryotic translation initiation factor 4E-binding protein 1 phosphorylated at Ser83. It has been testedfor use in Flow Cytometry, Immunocytochemistry, Immunofluorescence, & Western Blotting.
Immunohistochemistry Analysis: A 1:200 dilution from a representative lot detected phospho 4E-BP1 (Ser83) in FFPE MCC tissue (Courtesy of Dr. Yuan Chang at University of Pittsburgh).

Flow Cytometry Analysis: A representative lot detected phospho 4E-BP1 (Ser83) in Nocodazole-arrested HeLa cells (Velasquez, C., et. al. (2016). Proc Natl Acad Sci USA. 113(30):8466-71).

Immunocytochemistry Analysis: A representative lot detected phospho 4E-BP1 (Ser83) in HEK293 and U2OS cells (Velasquez, C., et. al. (2016). Proc Natl Acad Sci USA. 113(30):8466-71).

Western Blotting Analysis: A representative lot detected phospho 4E-BP1 (Ser83) in Nocodazole-arrested HeLa cells, GST-4E-BP1 treated with mitotic HeLa lysate (Velasquez, C., et. al. (2016). Proc Natl Acad Sci USA. 113(30):8466-71).

Western Blotting Analysis: A representative lot detected phospho 4E-BP1 (Ser83) in Hela cells treated with Nocodazole (Courtesy of Dr. Yuan Chang at University of Pittsburgh).

Immunofluorescence Analysis: A 1:500 dilution from a representative lot detected phospho 4E-BP1 (Ser83) in FFPE MCC tissue (Courtesy of Dr. Yuan Chang at University of Pittsburgh).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa cells treated with Nocodazole.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected phospho 4E-BP1 (Ser83) in lysate from HeLa cells treated with Nocodazole (100 ng/mL).

Target description

~19 kDa observed; 12.58 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Rabbit polyclonal antiserum with 0.02% sodium azide and 50% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitotic CDK1 and 4E-BP1 I: Loss of 4E-BP1 serine 82 phosphorylation promotes proliferative polycystic disease and lymphoma in aged or sublethally irradiated mice.
Sun, et al.
PLoS ONE, 18, e0282722-e0282722 (2023)
Rui Sun et al.
The Journal of biological chemistry, 294(31), 11840-11852 (2019-06-16)
Eukaryotic translation initiation factor 4E (eIF4E)-binding protein 1 (4E-BP1) inhibits cap-dependent translation in eukaryotes by competing with eIF4G for an interaction with eIF4E. Phosphorylation at Ser-83 of 4E-BP1 occurs during mitosis through the activity of cyclin-dependent kinase 1 (CDK1)/cyclin B

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service