Skip to Content
Merck
All Photos(1)

Key Documents

FTPF00789

Millipore

Silicone 4 in. Tri-Clover gasket 1-cartridge housing

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€79.00

€79.00


In StockDetails



Select a Size

Change View
1 EA
€79.00

About This Item

UNSPSC Code:
41104008
eCl@ss:
25140604

€79.00


In StockDetails


input

sample type pharmaceutical(s)

Quality Level

application(s)

environmental

compatibility

for use with M AIR T Compressed Gas and Air Test Kit

Legal Information

M AIR T is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service