Skip to Content
Merck
All Photos(1)

Key Documents

ABE284

Sigma-Aldrich

Anti-T-box transcription factor TBX5 Antibody

serum, from rabbit

Synonym(s):

T-box transcription factor TBX5, T-box protein 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€381.00

€381.00


Estimated to ship on26 March 2026Details


Request a Bulk Order

Select a Size

Change View
100 μL
€381.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€381.00


Estimated to ship on26 March 2026Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TBX5(6910)

General description

The gene encoding T-box transcription factor 5 (TBX5) is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene is closely linked to related family member T-box 3 (ulnar mammary syndrome) on human chromosome 12. The encoded protein may play a role in heart development and specification of limb identity.

Immunogen

Recombinant protein corresponding to mouse T-box transcription factor TBX5.

Application

Anti-T-box transcription factor TBX5 Antibody detects level of T-box transcription factor TBX5 & has been published & validated for use in Western Blotting, ICC & IHC.
Immunocytochemistry Analysis: A representative lot from an independent laboratory detected T-box transcription factor TBX5 in 293T cells overexpressing T-box transcription factor TBX5, mouse primary cardiocytes treated with serum, HA-tagged Tbx5 transfected NIH/3T3 cells, and HA-tagged Tbx5 transfected C2C12 cells (Georges, R., et al. (2008). Mol Cell Biol. 28(12):4052-4067.)

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected T-box transcription factor TBX5 in mouse embryonic tissue and in Tbx5 transgenic mouse heart tissues treated with tamoxifin (Georges, R., et al. (2008). Mol Cell Biol. 28(12):4052-4067.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: A 1:1,000 dilution from a representative lot detected T-box transcription factor TBX5 in 10 µg of HeLa cell lysate.

Target description

~53 kDa observed. An uncharacterized band may be observed at ~75 kDa in some cell lysates. Three isoforms due to alternative splicing may be observed at ~35 kDa, ~50 kDa, and ~64 kDa in some cell lysates. (Georges, R., et al. (2008). Mol Cell Biol. 28(12):4052-4067.).

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service