Skip to Content
Merck
All Photos(2)

Key Documents

ABE1418

Sigma-Aldrich

Anti-Histone acetyltransferase KAT2A

from rabbit, purified by affinity chromatography

Synonym(s):

General control of amino acid synthesis protein 5-like 2, Histone acetyltransferase GCN5, HsGCN5, Lysine acetyltransferase 2A, STAF97

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€130.00
100 μG
€317.00

€130.00


Estimated to ship on28 May 2025Details


Request a Bulk Order

Select a Size

Change View
25 μG
€130.00
100 μG
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€130.00


Estimated to ship on28 May 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

chicken (based on 100% sequence homology), rat (based on 100% sequence homology), bovine (based on 100% sequence homology), mouse (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... KAT2A(2648)

General description

Histone acetyltransferase KAT2A (UniProt: Q92830; also known as EC 2.3.1.48, General control of amino acid synthesis protein 5-like 2, Histone acetyltransferase GCN5, HsGCN5, Lysine acetyltransferase 2A, STAF97) is encoded by the KAT2A (also known as GCN5, GCN5L2, HGCN5) gene (Gene ID: 2648) in human. Histone acetyltransferases (HATs) are transcription co-activators recruited by transcriptional activators to target gene promoters, where they enhance the transcription activity by acetylating lysine residues within the N-terminal regions of histones. KAT2A has significant histone acetyltransferase activity with core histones, but not with nucleosome core particles. It is also shown to acetylate non-histone proteins, such as CEBPB. It is also responsible for the acetylation of histone H3 at the gamma-H2A.X harboring nucleosomes at the site of DNA damage, thus establishing a cross-talk between the two histone modifications. It is expressed in all tissues and high expression is observed in ovary. Two isoforms of KAT2A have been described that are produced by alternative splicing. It has three acetyl CoA-binding sites (aa 579-581; 586-592, and 617-620) and an N-acetyltransferase domain (aa 503-656) and a bromo domain (aa 745-815). Histone acetyltransferase KAT2A plays important roles in multiple stages of T cell functions, including development, clonal expansion, and differentiation. Loss of its activity is shown to impair T cell proliferation, IL-2 production, and Th1/Th17, but not Th2 and regulatory T cell differentiation. In case of HIV-1 infection, it is recruited by the viral protein Tat and regulates Tat′s transactivating activity and may help inducing chromatin remodeling of proviral genes. (Ref.: (Ref.: Gao, B et al. (2017). J. Immunol. 198(10); 3927-3938).

Specificity

This rabbit polyclonal antibody detects Histone acetyltransferase KAT2A in human cells. It targets an epitope within 15 amino acids from the C-terminal half.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal half of human Histone acetyltransferase KAT2A.

Application

Anti-Histone acetyltransferase KAT2A, Cat. No. ABE1418, is a highly specific rabbit polyclonal antibody that detects KAT2A (GCN5) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Histone acetyltransferase KAT2A in human pancreas, cerebral cortex, and tonsil tissue sections.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Histone acetyltransferase KAT2A in 10 µg of A431 cell lysate.

Target description

~94 kDa observed; 93.93 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service