Skip to Content
Merck
All Photos(2)

Key Documents

ABE1330

Sigma-Aldrich

Anti-FUSE-binding protein 1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Far upstream element-binding protein 1, FBP, FUSE-binding protein 1, DNA helicase V, hDH V

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€388.00

€388.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€388.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€388.00


Estimated to ship on08 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

feline (based on 100% sequence homology), primate (based on 100% sequence homology), rat (based on 100% sequence homology), mouse (based on 100% sequence homology), horse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FUBP1(8880)

General description

FUSE-binding protein 1, also known as Far upstream element-binding protein 1 FBP, FUSE-binding protein 1, DNA helicase V, hDH V, and encoded by the gene name FUBP1, regulates MYC expression by binding to a single-stranded far-upstream element (FUSE) upstream of the MYC promoter. FUSE may act both as activator and repressor of transcription. There are three known FUSE binding proteins (FBP1, FBP2, and FBP3). All three belong to an ancient family of single-stranded DNA binding proteins. These proteins are required for regulation of the c-myc proto-oncogene and have been shown to bind to an A/T-rich element (far-ustream element) located 1.7 kb upstream of the c-myc P2 promoter. Although their primary sequences are closely related, the FBP, FBP2, and FBP3 genes are located on different chromosomes in both mice and humans. FUSE binding proteins are believed to be multifunctional. Besides regulating the transcription of c-myc (and quite likely other targets), the FBP family has been shown to bind a variety of RNAs in vitro.

Immunogen

Epitope: Near N-terminal
KLH-conjugated linear peptide corresponding to human FUSE-binding protein 1 near the N-terminal.

Application

Immunocytochemistry Analysis: 2.0 µg/mL from a representative lot detected FUSE-binding protein 1 in HeLa and A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-FUSE-binding protein 1 | ABE1330 antibody is validated for use in WB, IC for the detection of FUSE-binding protein 1 | ABE1330.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected FUSE-binding protein 1 in 10 µg of HeLa cell lysate.

Target description

~76 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caiqi Ma et al.
Life sciences, 269, 119098-119098 (2021-01-22)
Far upstream element-binding protein 1 (FUBP1) has been shown to involve in the tumorigenesis and tumor progression of various cancers. However, the expression and function of FUBP1 in cervical carcinoma remains unknown. Transcriptional expression of FUBP1 was initially evaluated using
Ping Jiang et al.
Journal of experimental & clinical cancer research : CR, 38(1), 400-400 (2019-09-13)
Neuroblastoma (NB) is one of the deadliest paediatric solid tumours due to its rapid proliferative characteristics. Amplified copies of MYCN are considered the most important marker for the prediction of tumour relapse and progression in NB, but they were only
Haofan Yin et al.
Molecular oncology, 15(12), 3490-3512 (2021-07-22)
Distant metastasis is, unfortunately, the leading cause of death in colorectal cancer (CRC). Approximately 50% of CRC patients develop liver metastases, while 10-30% of patients develop pulmonary metastases. The occurrence of metastasis is considered to be almost exclusively driven by
Jingxin Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(20), E4604-E4612 (2018-05-02)
RG-7916 is a first-in-class drug candidate for the treatment of spinal muscular atrophy (SMA) that functions by modulating pre-mRNA splicing of the SMN2 gene, resulting in a 2.5-fold increase in survival of motor neuron (SMN) protein level, a key protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service