Skip to Content
Merck
All Photos(1)

Key Documents

ABE1025

Sigma-Aldrich

Anti-Achaete-scute homolog 1 Antibody (ASCL1)

from rabbit, purified by affinity chromatography

Synonym(s):

Achaete-scute homolog 1, ASH-1, hASH1, Class A basic helix-loop-helix protein 46, bHLHa46

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€356.00

€356.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€356.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€356.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ASCL1(429)

General description

Achaete-scute homolog 1 (ASCL1), also known as MASH1, ASH1 or HASH1, is a basic helix-loop-helix transcription factor. ASCL1 binds to the E box (5′-CANNTG-3′) which then activates transcription. For efficient DNA binding, dimerization with other BHLH proteins is required. Achaete Scute proteins may also be involved in the determination of neuronal precursors in the central and peripheral nervous systems. It is expressed in fetal brain and is essential for the proper development and the survival of subsets of autonomic neurons. It is highly expressed in medullary thyroid cancer and small cell lung cancer.

Immunogen

BCP-conjugated linear peptide corresponding to human Achaete-scute homolog 1 (ASCL1).

Application

This Anti-Achaete-scute homolog 1 Antibody (ASCL1) is validated for use in Western Blotting and Immunohistochemistry for the detection of Achaete-scute homolog 1 (ASCL1).
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected Achaete-scute homolog 1 (ASCL1) in 5 µg of mouse E13-14 brain lysate and A549 cell lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Achaete-scute homolog 1 (ASCL1) in rat cerebellum, rat cerebral cortex, and rat hippocampus tissue.

Quality

Evaluated by Western Blotting using lysate from cells over expressing Achaete-scute homolog 1 (ASCL1).

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Achaete-scute homolog 1 (ASCL1) in 5 µg of lysate from cells over expressing Achaete-scute homolog 1 (ASCL1).

Target description

~38 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satyanarayana Alleboina et al.
Molecular therapy oncolytics, 23, 181-191 (2021-11-04)
Retinoic acids (RAs) are the most successful therapeutics for cancer differentiation therapy used in high-risk neuroblastoma (NB) maintenance therapy but are limited in effectiveness. This study identifies a strategy for improving efficacy through disruption of cancer cell identity via BET

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service