Skip to Content
Merck
All Photos(3)

Key Documents

ABC462

Sigma-Aldrich

Anti-DNA polymerase subunit gamma-1 Antibody/POLG

1.0 mg/mL, from rabbit

Synonym(s):

DNA polymerase subunit gamma-1, DNA polymerase subunit gamma-1, PolG-alpha

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€379.00

€379.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€379.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€379.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POLG(5428)

General description

POLG is a mitochondrial protein that plays a major role in mitochondrial DNA replication and only for mitochondrial DNA. POLG is the catalytic subunit of a multiple protein complex that makes up the holo-enzyme that is the mitochondrial specific DNA polymerase gamma. POLG makes the alpha subunit of the complex. A number of diseases are associated with mutations that affect the function of POLG and the mitochondria including Leigh syndrome, various neurodegeneration diseases such as Parkinson’s and Alzheimer′s, as well as various skeletal muscle and ophthalmoplegia disorders and even male infertility. The polymerase that POLG ultimately comprises is Polymerase gamma (Pol γ) and is the only DNA polymerase that is active in mitochondria and that can replicate mtDNA.

Immunogen

KLH-conjugated linear peptide corresponding to human DNA polymerase subunit gamma-1/POLG near the N-terminus.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected DNA polymerase subunit gamma-1/POLG in human cerebellum, human cerebral cortex, human tonsil and human thalamus tissue.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated DNA polymerase subunit gamma-1/POLG in 0.5 mg of HeLa cell lysate.
This Anti-DNA polymerase subunit gamma-1 Antibody/POLG is validated for use in Immunohistochemistry and Immunoprecipitation for the detection of DNA polymerase subunit gamma-1/POLG.

Quality

Evaluated by Immunohistochemistry in human tonsil tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected DNA polymerase subunit gamma-1/POLG in human tonsil tissue.

Target description

~140 kDa observed. Uncharacterized band(s) may appear in some lysates.

Linkage

Replaces: MABN1088

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel R Crooks et al.
Science signaling, 14(664) (2021-01-07)
Understanding the mechanisms of the Warburg shift to aerobic glycolysis is critical to defining the metabolic basis of cancer. Hereditary leiomyomatosis and renal cell carcinoma (HLRCC) is an aggressive cancer characterized by biallelic inactivation of the gene encoding the Krebs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service