Skip to Content
Merck
All Photos(2)

Key Documents

ABC157

Sigma-Aldrich

Anti-MCM7 Antibody

from rabbit

Synonym(s):

DNA replication licensing factor MCM7, CDC47 homolog, P1.1-MCM3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€344.00

€344.00


Estimated to ship on17 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€344.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€344.00


Estimated to ship on17 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MCM7(4176)

General description

MCM7 is a highly conserved mini-chromosome maintenance protein (MCM) that is involved in the initiation of eukaryotic genome replication. It forms a hexameric complex with MCM4 and MCM6 (two subunits of MCM7, two subunits of MCM4 and two subunits of MCM7) and this complex regulates the helicase activity of the pre-replication complex. This complex is also involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. MCM7 is reported to bind to DNA in metaphase II in oocytes and remains associated with the DNA until the S-phase. Two isoforms of MCM7 have been reported, 61 kDa and 90 kDa. In neuroblastoma tumors MCM7 expression is shown to correlates with MYCN (v-myc myelocytomatosis viral related oncogene, neuroblastoma derived) amplification where in MCM7 is rapidly up-regulated by Inducible MYCN Expression. MYCN protein is shown to bind specifically to the MCM7 promoter region, which possesses an E-box binding site for the Myc oncoproteins.

Specificity

Other homologies: Mouse (71% sequence homology).
This antibody recognizes the N-terminus of MCM7

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human MCM7.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This MCM7 antibody is validated for use in WB for the detection of the MCM7 protein.
Western Blot Analysis: 0.5 µg/mL from a representative lot detected MCM7 in 10 µg of L6 cell lysate.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected MCM7 in 10 µg of HeLa cell lysate.

Target description

~82 kDa observed

Linkage

Replaces: MABE188

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service