Skip to Content
Merck
All Photos(1)

Key Documents

ABC139

Sigma-Aldrich

Anti-TSPO Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Translocator protein, Mitochondrial benzodiazepine receptor, PKBS, Peripheral-type benzodiazepine receptor, PBR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€390.00

€390.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€390.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€390.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TSPO(706)

General description

Translocator protein (TSPO) is also called Mitochondrial benzodiazepine receptor, PKBS, or Peripheral-type benzodiazepine receptor (PBR). TSPO generates peripheral-type benzodiazepine recognition sites. TSPO has a role in cholesterol transport across mitochondrial membranes in steroidogenic cells, and may also be involved in in the transport of porphyrins and heme. TSPO is found in many tissue types, but is normally found expressed at the highest levels in steroid synthesizing tissues.

Specificity

This antibody recognizes the C-terminus of TSPO.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human TSPO.

Application

Anti-TSPO Antibody is a rabbit polyclonal antibody, validated for use in western blotting & IHC.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected TSPO in 10 µg of HeLa cell lysate.
Immunohistochemistry Analysis: A 1:500-2,000 dilution from a representative lot detected TSPO in human thalamus, human cardiac myocytes and human cerebellum tissue.

Quality

Evaluated by Western Blotting in human testis tissue lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected TSPO in 10 µg of human testis tissue lysate.

Target description

~16 kDa observed

Linkage

Replaces: MABC650

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human testis tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faheem Ullah et al.
Frontiers in neuroscience, 14, 170-170 (2020-04-01)
Chronic glial activation is characterized by an increased number of activated microglia and astroglia; these secrete free radicals and cytotoxic cytokines, subsequently causing neuronal damage. This study investigated the hypothesis that a soy-lecithin based phytosomal curcumin formulation can decrease glial
Faheem Ullah et al.
Scientific reports, 10(1), 2365-2365 (2020-02-13)
Chronic glial activation is characterized by increased numbers of activated glial cells, secreting free radicals and cytotoxic cytokines, subsequently causing neuronal damage. In order to investigate the anti-inflammatory activity of Longvida® Optimised Curcumin (LC), we fed 500 ppm of LC to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service