Skip to Content
Merck
All Photos(1)

Key Documents

SML0729

Sigma-Aldrich

BAM8-22 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

BAM (8-22) trifluoroacetate salt, Bovine Adrenal Medulla 8-22 trifluoroacetate salt, Bovine adrenal medulla peptide (8-22) trifluoroacetate salt, L-Valyl-glycyl-L-arginyl-L-prolyl-L-α-glutamyl-L-tryptophyl-L-tryptophyl-L-methionyl-L-α-aspartyl-L-tyrosyl-L-glutaminyl-L-lysyl-L-arginyl-L-tyrosyl-glycine trifluoroacetate salt, VGRPEWWMDYQKRYG trifluoroacetate salt, Val-Gly-Arg-Pro-Glu-Trp-Trp-Met-Asp-Tyr-Gln-Lys-Arg-Tyr-Gly trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€440.00

€440.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€440.00

About This Item

Empirical Formula (Hill Notation):
C91H127N25O23S · xC2HF3O2
CAS Number:
Molecular Weight:
1971.20 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€440.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

S(CC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H]7N(CCC7)C(=O)[C@@H](NC(=O)CNC(=O)[C@@H](N)C(C)C)CCCN\C(=N\[H])\N)CCC(=O)O)Cc5c6c([nH]c5)cccc6)Cc3c4c([nH]c3)cccc4)C(=O)N[C@@H](CC(=O)O)C(=O)N[C@@H](Cc2ccc(cc2)O)C(=O)N[C@@H](CCC(=O)N)C(=O)N[C@@H]

InChI

1S/C91H127N25O23S/c1-48(2)76(94)88(138)103-46-72(120)105-64(19-11-36-100-91(97)98)89(139)116-37-12-20-70(116)87(137)110-62(30-32-73(121)122)81(131)113-68(42-52-45-102-58-16-7-5-14-56(52)58)85(135)114-67(41-51-44-101-57-15-6-4-13-55(51)57)84(134)109-63(33-38-140-3)82(132)115-69(43-74(123)124)86(136)112-66(40-50-23-27-54(118)28-24-50)83(133)108-61(29-31-71(93)119)80(130)106-59(17-8-9-34-92)78(128)107-60(18-10-35-99-90(95)96)79(129)111-65(77(127)104-47-75(125)126)39-49-21-25-53(117)26-22-49/h4-7,13-16,21-28,44-45,48,59-70,76,101-102,117-118H,8-12,17-20,29-43,46-47,92,94H2,1-3H3,(H2,93,119)(H,103,138)(H,104,127)(H,105,120)(H,106,130)(H,107,128)(H,108,133)(H,109,134)(H,110,137)(H,111,129)(H,112,136)(H,113,131)(H,114,135)(H,115,132)(H,121,122)(H,123,124)(H,125,126)(H4,95,96,99)(H4,97,98,100)/t59-,60-,61-,62-,63-,64-,65-,66-,67-,68-,69-,70-,76-/m0/s1

InChI key

GOEYECACIBFJGZ-NPAGUKBMSA-N

Amino Acid Sequence

Val-Gly-Arg-Pro-Glu-Trp-Trp-Met-Asp-Tyr-Gln-Lys-Arg-Tyr-Gly trifluoroacetate salt

Application

BAM8-22 trifluoroacetate salt has been used in the study to identify the functions of Mas-related gene (Mrg) C in the pathogenesis and treatment of bone cancer pain (BCP).[1]

Biochem/physiol Actions

BAM8-22 is a selective agonist for the sensory neuron specific G-protein coupled receptors (SNSRs).
BAM8-22 is a selective agonist for the sensory neuron specific G-protein coupled receptors (SNSRs). BAM8-22 inhibits development of morphine tolerance in rats. BAM8-22 is inactive at opioid receptors. Also, BAM8–22 is a potent activator of Mas-related G-protein-coupled receptors (Mrgprs), MrgprC11 and hMrgprX1. BAM8-22 produces itch and nociceptive sensations in humans in a histamine-independent manner.
Bovine adrenal medulla 8-22 (BAM8-22), a 15-amino acid peptide, acts a cleavage product of proenkephalin A and the precursor of Leu- and Met-enkephalin in the adrenal medulla. It may also be used to treat pain. It blocks inflammatory pain, chemical pain and downregulates the expression of spinal c-fos gene in an opioid-independent manner. In mice, BAM8-22 diminishes bone cancer pain. It induces the expression of Mas-related G-protein-coupled receptor C (MrgC) in the spinal dorsal horn.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mas-related G-protein-coupled receptor c agonist bovine adrenal medulla 8-22 attenuates bone cancer pain in mice.
Sun YE, et al.
International Journal of Clinical and Experimental Medicine, 8(11), 20178-20178 (2015)
Mas-Related Gene (Mrg) C Activation Attenuates Bone Cancer Pain via Modulating Gi and NR2B.
Sun YE, et al.
PLoS ONE, 11(5), e0154851-e0154851 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service