Skip to Content
Merck
All Photos(2)

Key Documents

AB9728

Sigma-Aldrich

Anti-S100A12 Antibody

Chemicon®, from rabbit

Synonym(s):

EN-RAGE, Extacellular newly identified RAGE-binding protein, CAAF1, Calcium-binding protein in amniotic fluid-1, Calgranulin C, Calgranulin-related protein, CGRP, Cornea-associated antigen, CO-Ag

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€497.00

€497.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€497.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€497.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... S100A12(6283)

General description

S100 calcium binding protein A12, also known as S100A12, EN-RAGE, and calgranulin C, is a proinflammatory protein with significant potential as a biomarker. It is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. The member S100A12 is proposed to be involved in specific calcium-dependent signal transduction pathways and its regulatory effect on cytoskeletal components may modulate various neutrophil activities. It has been reported to be secreted by activated granulocytes and binds to the receptor for advanced gycation end products, which induces NfkappaB-dependent activation of endothelium (Foell et al., 2003). S100A12 is strongly expressed in inflamed synovial tissue typical in rheumatoid and psoriatic arthritis and may also prove an excellent biomarker for Inflammatory bowel disease (Kaiser et al., 2007). S100A12 protein may also be involved in chronic inflammation in atherosclerotic lesions including those of Type II diabetics (Kosaki et al. 2004) and the acceleration of atherosclerosis in hemodialysis patients (Mori et al., 2009).

Foell, D. et al. (2003). Expression of the pro-inflammatory protein S100A12 (EN-RAGE) in rheumatoid and psoriatic arthritis. Rheumatol. 42:1383-1389.
Kaiser, T. et al. (2007). Faecal S100A12 as a non-invasive marker distinguishing inflammatory bowel disease from irritable bowel syndrome. Gut. 56:1706-1713.
Kosaki, A. et al. (2004). Increased plasma S100A12 (EN-RAGE) levels in patients with type 2 diabetes. J. Clin. Endocrinol. Metabol. 89:5423-5428.
Mori, Y. et al. (2009). Increased Plasma S100A12 (EN-RAGE) levels in hemodialysis patients with atherosclerosis. Am. J. Nephrol. 29:18-24.

Specificity

By western blot this polyclonal antibody recognizes a band at ~10-11 kDa under reducing conditions. A dimer can be sometimes be seen at ~ 21 kDa using recombinant human S100A12. Antibody was affinity purifed over immobilized S100A12 prior to vialing.

Immunogen

Full length recombinant His-tagged human S100A12 that was expressed in E. coli and purified by Ni-NTA affinity chromatography prior to immunization.

Application

Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Neuroinflammation & Pain
This Anti-S100A12 Antibody is validated for use in ELISA, IH, WB for the detection of S100A12.
Western blot: 1:1,000-1:10,000 on recombinant human S100A12.
Immunohistochemistry: fesh frozen, acetone fixed sections 1:100-1:500 using ABC enzymatic detection methods (Srikrishna, G. et al, 2005). Other fixation conditions not yet tested.
ELISA: 1:10,000-1:100,000 against the recombinant human S100A12.
It is expected that the antibody will also work for immunohistochemistry.

Optimal working dilutions must be determined by the end user.

Physical form

Liquid in PBS with 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
HL-60 cells, and human PMN/Granulocytes {Vogl, T et al (1999) “S100A12 Is Expressed Exclusively by Granulocytes and Acts Independently from MRP8 and MRP14” J. Biol. Chem. 274(3):25291-25296}.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service