Skip to Content
Merck
All Photos(1)

Documents

P7505

Sigma-Aldrich

Putrescine dihydrochloride

≥98% (TLC), powder

Synonym(s):

1,4-Butanediamine dihydrochloride, 1,4-Diaminobutane dihydrochloride, Tetramethylenediamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4NH2 · 2HCl
CAS Number:
Molecular Weight:
161.07
Beilstein:
3906680
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Putrescine dihydrochloride, ≥98% (TLC)

Assay

≥98% (TLC)

form

powder

mp

280 °C (dec.) (lit.)

solubility

H2O: soluble 100 mg/mL, clear, colorless

SMILES string

Cl[H].Cl[H].NCCCCN

InChI

1S/C4H12N2.2ClH/c5-3-1-2-4-6;;/h1-6H2;2*1H

InChI key

XXWCODXIQWIHQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Putrescine dihydrochloride has been used:
  • as a bioactive amine standard in high-performance liquid chromatography (HPLC)
  • as a component of hormone mix supplemented to Dulbecco′s modified eagle medium (DMEM) to study its effects on neural stem cells
  • as a supplement in DMEM for culturing of endometrial cancer cells

Biochem/physiol Actions

Binds to the polyamine modulatory site of the NMDA receptor and potentiates NMDA-induced currents; precursor of spermidine.
Putrescine is an aliphatic amine and is positively charged. It acts as a precursor of spermine and plays a role in polyamine biosynthesis. Putrescine is found in several plants, animals, and bacteria. It plays a role in cell proliferation and cell growth in Escherichia coli. Putrescine is a component of agrochemicals, surfactants, pharmaceuticals, polymers, and additives.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bangmin Liu et al.
Journal of animal science and biotechnology, 10, 69-69 (2019-09-14)
Polyamines are essential for cell growth and beneficial for intestinal maturation. To evaluate the effects of putrescine on alleviating intestinal atrophy and underlying molecular mechanisms, both in vivo feeding trial and in vitro cell culture were conducted. Weanling pigs were
Ulises Gómez-Pinedo et al.
Frontiers in neurology, 8, 255-255 (2017-06-22)
Alexander disease (AxD) is a rare disease caused by mutations in the gene encoding glial fibrillary acidic protein (GFAP). The disease is characterized by presence of GFAP aggregates in the cytoplasm of astrocytes and loss of myelin. Determine the effect
Mito Kanatsu-Shinohara et al.
Methods in enzymology, 477, 17-36 (2010-08-12)
Spermatogonial stem cells (SSCs) in the testes are a new target for germline modification. With the development of an in vitro culture system and spermatogonial transplantation technique, SSCs can now be manipulated and used as an alternative to embryonic stem
Zhi-Gang Qian et al.
Biotechnology and bioengineering, 104(4), 651-662 (2009-08-29)
A four carbon linear chain diamine, putrescine (1,4-diaminobutane), is an important platform chemical having a wide range of applications in chemical industry. Biotechnological production of putrescine from renewable feedstock is a promising alternative to the chemical synthesis that originates from
Vegetables consumed in Brazilian cuisine as sources of bioactive amines
Dala-Paula B M, et al.
Food Bioscience, 40, 100856-100856 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service