Skip to Content
Merck
All Photos(1)

Key Documents

AB15668

Sigma-Aldrich

Anti-PYY Antibody, amino acids 3-18

Chemicon®, from chicken

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€287.00

€287.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€287.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€287.00


Estimated to ship on02 May 2025Details


Request a Bulk Order

biological source

chicken

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PYY(5697)
mouse ... Pyy(217212)

Specificity

PYY, amino acids 3-18.

Immunogen

Synthetic peptide from the N-terminus of human PYY (amino acids 3-18).

Application

Detect PYY using this Anti-PYY Antibody, amino acids 3-18 validated for use in ELISA.
ELISA: 1:200-1:1,000 when using as the coating antibody and 1:2,000-1:5,000 when using as the detection antibody in a sandwich ELISA.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Physical form

Liquid in PBS with 0.02% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service