Skip to Content
Merck
All Photos(1)

Documents

9Q071S

Sigma-Aldrich

Anti-Chlamydia LPS MAb Antibody, clone 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

affinity purified immunoglobulin

clone

clone 2
monoclonal

purified by

affinity chromatography

species reactivity

mouse, all

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
immunodiffusion: suitable
western blot: suitable

isotype

IgG2b

General description

Chlamydia Trachomatis LPS

Specificity

Specific for S. paratyphi

Application

Recommended pair for double antibody immunoassay: Coating: MABX8492; labeling: MABX8491

Physical form

10 mM PBS pH 7.0 Phosphate, with 0.05% NaN3

Storage and Stability

2-8°C. For long-term storage, freeze at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service