Skip to Content
Merck
All Photos(1)

Key Documents

AB1120F

Sigma-Aldrich

Anti-Chlamydia trachomatis-EB Antibody, FITC-conjugated

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€468.00

€468.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
€468.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€468.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

Specificity

Reacts with purified elementary bodies, disrupted. The unconjugated antibody reacts at >1:1,000 by indirect immunofluorescence vs. all serovars (A-K, L1-L3). Cross-reacts with Chlamydia psittacii and Chlamydia pneumoniae (TWAR). Non reactive with HEp-2 cells and egg yolk sac.

Immunogen

L2 and other serovar groups

Application

Applications include direct FA staining of target antigens in a permissive tissue culture system. Acetone fixation of the antigen source is recommended prior to staining.

Titer by indirect immunofluorescence: >1:1,000 vs. EB′s (all serovars, A-K, L1-L3).

Working dilutions must be determined by end user (suggested starting range 1:10-1:50).
Detect Chlamydia trachomatis-EB using this Anti-Chlamydia trachomatis-EB Antibody, FITC-conjugated validated for use in IF.

Physical form

IgG fraction conjugated with FITC. Supplied in PBS (0.1 M, pH 7.2) with 10 mg/mL BSA as a stabilizer and 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months or at 2-8°C for up to 3 months after date of receipt. Avoid repeated freeze/thaw cycles. Store under subdued light.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph S Park et al.
iScience, 11, 71-84 (2018-12-28)
The invasion of Chlamydia trachomatis, an obligate intracellular bacterium, into epithelial cells is driven by a complex interplay of host and bacterial factors. To comprehensively define the host genes required for pathogen invasion, we undertook a fluorescence-activated cell sorting (FACS)-based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service