Skip to Content
Merck
All Photos(1)

Key Documents

9630-OP

Sigma-Aldrich

X-Gal

OmniPur® Grade

Synonym(s):

OmniPur® X-Gal, 5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, BCIG, X-Gal

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€172.00

€172.00


Estimated to ship on28 April 2025Details



Select a Size

Change View
1 G
€172.00

About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

€172.00


Estimated to ship on28 April 2025Details


Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

shipped in

ambient

SMILES string

[Cu+2].[Cu+].N3(CCN(CC3)C)Cc1c(c(cc(c1)C#N)CN2CCN(CC2)C)O.OC(O)C.OC(O)C

InChI

1S/C19H29N5O.2C2H6O2.2Cu/c1-21-3-7-23(8-4-21)14-17-11-16(13-20)12-18(19(17)25)15-24-9-5-22(2)6-10-24;2*1-2(3)4;;/h11-12,25H,3-10,14-15H2,1-2H3;2*2-4H,1H3;;/q;;;+1;+2

InChI key

GGILAJWFDJWIQO-UHFFFAOYSA-N

General description

X-Gal is a chromogenic substrate used in conjunction with IPTG for blue/white colony selection.[1] It is a soluble colorless compound comprising galactose attached to a substituted indole.[2] In the presence of X-Gal the lacbacteria after α-complementation form blue colonies and bacteria carrying recombinant plasmid form white colonies.[1]

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Michael R Green et al.
Cold Spring Harbor protocols, 2019(12) (2019-12-04)
Many plasmid vectors (e.g., the pUC series, Bluescript, pGem, and their derivatives) carry a short segment of Escherichia coli DNA containing the regulatory sequences and the coding information for the first 146 amino acids of β-galactosidase. Vectors of this type
Douglas H Juers et al.
Protein science : a publication of the Protein Society, 21(12), 1792-1807 (2012-09-27)
This review provides an overview of the structure, function, and catalytic mechanism of lacZ β-galactosidase. The protein played a central role in Jacob and Monod's development of the operon model for the regulation of gene expression. Determination of the crystal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service