Skip to Content
Merck
All Photos(1)

Key Documents

8750-OP

Millipore

TAE Buffer

OmniPur® Grade, 25X, Premixed Powder

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
Inquire


Estimated to ship on09 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 PKG
Inquire

About This Item

UNSPSC Code:
41105319
NACRES:
NA.28


Estimated to ship on09 April 2025Details


Request a Bulk Order

Quality Level

form

fine powder

manufacturer/tradename

Calbiochem®

color

white

shipped in

ambient

General description

Each pouch makes 1L of 25X TAE Buffer(1X = 0.04M TRIS, 0.04M Acetate, 0.001M EDTA). Molecular Biology grade.

Specifications:
DNase: None detected
pH at 25°C: 8.2-8.4
Protease: None detected
RNase: None detected
Solubility (10%, water): To pass test





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service