Skip to Content
Merck
All Photos(1)

Key Documents

8720-OP

Millipore

TAE Buffer

OmniPur® Grade, 10X, Liquid Concentrate

Synonym(s):

TRIS-Acetate-EDTA Buffer

Sign Into View Organizational & Contract Pricing

Select a Size

4 L
€145.00

€145.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
4 L
€145.00

About This Item

UNSPSC Code:
12161700
NACRES:
NA.28

€145.00


Estimated to ship on15 April 2025Details


Request a Bulk Order

Quality Level

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear to haze-free colorless

shipped in

ambient

storage temp.

10-30°C

General description



General Composition: Each liter of concentrate makes 10L of 1X TAE Buffer (1X = 0.04 M TRIS, 0.04 M Acetate, 0.001 M EDTA). Molecular Biology grade.

DNase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service