Skip to Content
Merck
All Photos(1)

Key Documents

8.52077

Sigma-Aldrich

Fmoc-Glu-ODmab

Novabiochem®

Synonym(s):

Fmoc-Glu-ODmab, N-α-Fmoc-L-glutamic acid α-4-{N-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl]-amino} benzyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€282.00
5 G
€1,120.00

€282.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€282.00
5 G
€1,120.00

About This Item

Empirical Formula (Hill Notation):
C40H44N2O8
CAS Number:
Molecular Weight:
680.79
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

€282.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥90% (TLC)
≥96.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

carboxylic acid

storage temp.

15-25°C

SMILES string

N(C(CCC(=O)OCc4ccc(cc4)NC(=C5C(=O)CC(CC5=O)(C)C)CC(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C40H44N2O8/c1-24(2)19-33(37-34(43)20-40(3,4)21-35(37)44)41-26-15-13-25(14-16-26)22-49-36(45)18-17-32(38(46)47)42-39(48)50-23-31-29-11-7-5-9-27(29)28-10-6-8-12-30(28)31/h5-16,24,31-32,41H,17-23H2,1-4H3,(H,42,48)(H,46,47)

InChI key

DABQNXPVTHIRRK-UHFFFAOYSA-N

General description

Quasi-orthogonally-protected Glu derivative. The Dmab group can be removed selectively in the presence of tBu-based protecting groups by treatment with 2% hydrazine in DMF [1], making this derivative an extremely useful tool for the preparation of cyclic peptides by Fmoc SPPS or for library synthesis. For the on-resin synthesis of side-chain to side-chain lactam bridged peptides, the combination of Lys(ivDde) and Glu(ODmab) is particularly advantageous since both side-chains can be simultaneously unmasked in a single step.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] W. C. Chan, et al. (1995) J. Chem. Soc., Chem. Commun., 2209.

Linkage

Replaces: 04-12-1174

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (TLC(011A)): ≥ 90 %
Purity (TLC(0811)): ≥ 90 %
Assay (HPLC, area%): ≥ 96.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service