Skip to Content
Merck
All Photos(1)

Key Documents

289647

Sigma-Aldrich

3-Amino-4-hydroxybenzoic acid

97%, for peptide synthesis

Synonym(s):

4-Hydroxy-3-aminobenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€30.30
25 G
€60.60

€30.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€30.30
25 G
€60.60

About This Item

Linear Formula:
H2NC6H3(OH)CO2H
CAS Number:
Molecular Weight:
153.14
Beilstein:
972238
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

€30.30


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

3-Amino-4-hydroxybenzoic acid, 97%

Quality Level

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

208 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Nc1cc(ccc1O)C(O)=O

InChI

1S/C7H7NO3/c8-5-3-4(7(10)11)1-2-6(5)9/h1-3,9H,8H2,(H,10,11)

InChI key

MRBKRZAPGUCWOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Gerecke et al.
Epigenetics, 15(3), 307-322 (2019-09-12)
Mutations in the enzyme isocitrate dehydrogenase 1 (IDH1) lead to metabolic alterations and a sustained formation of 2-hydroxyglutarate (2-HG). 2-HG is an oncometabolite as it inhibits the activity of α-ketoglutarate-dependent dioxygenases such as ten-eleven translocation (TET) enzymes. Inhibitors of mutant
Experience with cheetan in management of HIV infection.
J Chege et al.
East African medical journal, 74(7), 460-461 (1997-07-01)
Hirokazu Suzuki et al.
Journal of bacteriology, 189(5), 2155-2159 (2006-12-13)
An arylamine N-acetyltransferase (NAT) responsible for the N acetylation of exogenous 3-amino-4-hydroxybenzoic acid in Streptomyces griseus was identified and characterized. This enzyme was distinct from other eukaryotic and bacterial NATs in that it acetylated various 2-aminophenol derivatives more effectively than
Hirokazu Suzuki et al.
The Journal of biological chemistry, 281(48), 36944-36951 (2006-09-28)
The shikimate pathway, including seven enzymatic steps for production of chorismate via shikimate from phosphoenolpyruvate and erythrose-4-phosphate, is common in various organisms for the biosynthesis of not only aromatic amino acids but also most biogenic benzene derivatives. 3-Amino-4-hydroxybenzoic acid (3,4-AHBA)
Hirokazu Suzuki et al.
The Journal of antibiotics, 60(6), 380-387 (2007-07-10)
In grixazone biosynthesis by Streptomyces griseus, a key intermediate 3-amino-4-hydroxybenzoic acid (3,4-AHBA) is converted to another key intermediate 3-amino-4-hydroxybenzaldehyde (3,4-AHBAL). Two genes griC and griD in the grixazone biosynthesis gene cluster were found to be responsible for this conversion, because

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service