Skip to Content
Merck
All Photos(1)

Key Documents

8.20738

Sigma-Aldrich

Iodine monobromide

for synthesis

Synonym(s):

Iodine monobromide, Bromine iodide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€104.00
250 G
€220.00

€104.00


Estimated to ship on30 January 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€104.00
250 G
€220.00

About This Item

Empirical Formula (Hill Notation):
BrI
CAS Number:
Molecular Weight:
206.81
MDL number:
UNSPSC Code:
12352307
EC Index Number:
232-159-9
NACRES:
NA.22
form:
solid

€104.00


Estimated to ship on30 January 2025Details


Request a Bulk Order

Quality Level

form

solid

reaction suitability

reagent type: oxidant

bp

116 °C/1013 hPa (decomposition)

mp

37-40 °C

density

4.41 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

IBr

InChI

1S/BrI/c1-2

InChI key

CBEQRNSPHCCXSH-UHFFFAOYSA-N

Application

  • Graphite Intercalation Compounds: Explores the thermoelectric power of graphite intercalation compounds with iodine monobromide, enhancing understanding of material properties for electronic applications (Kobayashia and Sugiharaa, 2019).
  • Liquid-Crystalline Complexes: Studies halogen-bonded liquid-crystalline complexes formed from iodine and interhalogens like iodine monobromide, which are significant for developing new materials with adjustable properties (McAllister et al., 2023).

Analysis Note

Assay (iodometric): ≥ 95.0 %
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service