Skip to Content
Merck
All Photos(1)

Key Documents

1.09164

Supelco

Hanus solution

c(IBr) = 0.1 mol/l, for determination of iodine number , ready-to-use volumetric solution for titration, Titriplex®

Synonym(s):

Iodine solution acc. to Hanus

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€142.00
1 L
€149.00

€142.00


Estimated to ship on03 March 2025Details


Request more information

Select a Size

Change View
1 L
€142.00
1 L
€149.00

About This Item

UNSPSC Code:
12352402
NACRES:
NA.21

€142.00


Estimated to ship on03 March 2025Details


Request more information

Product Name

Hanus solution, for determination of iodine number c(IBr) = 0.1 mol/l, Titripur®

Quality Level

product line

Titripur®

form

liquid

quality

Analyzed in our ISO 17025 accredited QC lab

reaction suitability

reaction type: Redox Reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

<1 (20 °C in H2O)

transition temp

flash point 40 °C

density

1.06 g/cm3 at 20 °C

storage temp.

15-25°C

Features and Benefits

Accreditation:
This volumetric solution is analyzed by our calibration laboratory D-K-15185-01-00 which is accredited according to DIN EN ISO/IEC 17025 for analysis of amount-of-substance concentrations in volumetric solutions by DAkkS (Deutsche Akkreditierungsstelle - German National Accreditation Body). The accreditation certificate can be found at www.sigmaaldrich.com/ISO17025.

Packaging

SKU-pack size numbers ending with a "3" are SmartChemicals with an RFID tag on the label for seamless data transfer to the instrument, e.g. xxxxxx1003 or xxxxxxx 4003. More information on www.sigmaaldrich.com/SmartChemicals

Analysis Note

Form liquid
Amount-of-substance concentration 0.0994 - 1.006 mol/L
Measurement uncertainty ± 0.0004 mol/L
Traceability NIST SRM

The concentration is determined by volumetric titration and refers to 20°C.
The amount-of-substance concentration of this volumetric solution is determined with standardized sodium thiosulfate solution (article number 1.09147). The sodium thiosulfate solution is standardized and traceable to a primary standard reference material (SRM) from the National Institute of Standards and Technology, Gaithersburg, USA (NIST SRM 136 potassium dichromate) by means of volumetric standard potassium iodate (article number 1.02404), certified reference material according to ISO 17034, analyzed by our accredited calibration laboratory of Merck KGaA, Darmstadt, Germany according to DIN EN ISO/IEC 17025. The uncertainty is expressed as expanded measurement uncertainty with a coverage factor k=2 covering a confidence level of 95%.

Note: The titer is a correction factor to correct for variations of the volumetric solution, the titration equipment, the temperature and other laboratory conditions. For correct titration results it is recommended to determine a titer with the laboratory specific equipment and under laboratory specific conditions directly after opening a new bottle and at regular time intervals.

Legal Information

Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

SmartChemicals enable wireless CoA data transfer, reducing human error in titration experiments.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service