Skip to Content
Merck
All Photos(1)

Key Documents

8.20252

Sigma-Aldrich

Dibutyl sulfide

for synthesis

Synonym(s):

Dibutyl sulfide, Butyl sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€68.10
100 ML
€92.70
500 ML
€354.00
25 L
€16,040.00

€68.10


Estimated to ship on27 March 2025Details


Request a Bulk Order

Select a Size

Change View
50 ML
€68.10
100 ML
€92.70
500 ML
€354.00
25 L
€16,040.00

About This Item

Linear Formula:
CH3(CH2)3S(CH2)3CH3
CAS Number:
Molecular Weight:
146.29
MDL number:
UNSPSC Code:
12352113
EC Index Number:
208-870-5
NACRES:
NA.22

€68.10


Estimated to ship on27 March 2025Details


Request a Bulk Order

vapor pressure

26 hPa ( 80 °C)

Quality Level

Assay

≥96.0% (GC)

form

liquid

potency

2220 mg/kg LD50, oral (Rat)

bp

188.7 °C/1013 hPa

mp

-80 °C

transition temp

flash point 62 °C

density

0.84 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

S(CCCC)CCCC

InChI

1S/C8H18S/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3

InChI key

HTIRHQRTDBPHNZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.837 - 0.839
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service