Skip to Content
Merck
All Photos(1)

Documents

8.18505

Sigma-Aldrich

5-Fluorouracil

for synthesis

Synonym(s):

5-Fluorouracil, 5-Fluoro-2,4-dihydroxypyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3FN2O2
CAS Number:
Molecular Weight:
130.08
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-085-6
NACRES:
NA.22

Quality Level

form

powder

potency

230 mg/kg LD50, oral (Rat)

pH

4.3-5.3 (20 °C, 10 g/L in H2O)

bp

190-200 °C/0.13 hPa

mp

282-283 °C (decomposition)

storage temp.

2-30°C

InChI

1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)

InChI key

GHASVSINZRGABV-UHFFFAOYSA-N

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service