Skip to Content
Merck
All Photos(1)

Key Documents

8.20354

Sigma-Aldrich

Curcumin

for synthesis

Synonym(s):

Curcumin, 1,7-Bis(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Turmeric yellow, Diferuloylmethane

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
€53.30
10 G
€80.50
50 G
€264.00

€53.30


Estimated to ship on07 February 2025Details


Request a Bulk Order

Select a Size

Change View
2 G
€53.30
10 G
€80.50
50 G
€264.00

About This Item

Linear Formula:
[4-(OH)-3-(CH3O)C6H3CH=CHCO]2CH2
CAS Number:
Molecular Weight:
368.38
MDL number:
UNSPSC Code:
12352115
EC Index Number:
207-280-5
NACRES:
NA.22

€53.30


Estimated to ship on07 February 2025Details


Request a Bulk Order

Quality Level

form

powder

potency

>5000 mg/kg LD50, oral (Rat)

mp

170-175 °C

storage temp.

2-30°C

InChI

1S/C21H20O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h3-13,22,24-25H,1-2H3/b7-3+,8-4+,16-13-

InChI key

ZIUSSTSXXLLKKK-KOBPDPAPSA-N

Application

  • Enhancing Growth Performance in Livestock: A study demonstrated the use of a Curcumin/Silymarin/Yeast-based mycotoxin detoxifier to improve redox status and growth performance in weaned piglets, indicating curcumin′s potential application in animal health and nutrition (Papatsiros et al., 2024).
  • Drug Delivery in Biomedical Applications: Evaluation of a pH-responsive dextran-based silver nanocomposite hydrogel indicated significant potential in drug release efficiency and antibacterial property, offering a promising method for curcumin delivery in therapeutic settings (Luanda et al., 2024).
  • Anti-Inflammatory and Antioxidative Nanoparticles: A study developed Reactive Oxygen Species-responsive dual anti-inflammatory and antioxidative nanoparticles, suggesting applications for anterior uveitis treatment and showcasing curcumin′s pharmacological properties (Zhang et al., 2024).
  • Smartphone-based Detection of Food Freshness: A development in amine response smartphone-based portable and intelligent polyvinyl alcohol films provided real-time detection of shrimp freshness, representing an innovative industrial application of curcumin (Luo et al., 2024).

Analysis Note

Assay (HPLC, area%): ≥ 75.0 % (a/a)
Bisdemethoxycurcumin (HPLC; Area%): ≤ 5.0 % (a/a)
Demethoxycurcumin (HPLC; Area%): ≤ 20.0 % (a/a)
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service